Novus Biologicals
Manufacturer Code:NBP299845B
Catalog # NB089783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FLAP (NP_001620.2 Met1-Pro161). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Arachidonate 5-lipoxygenase-activating protein; arachidonate 5-lipoxygenase-activating protein FLAPMK-886-binding protein; FLAP; MK-886-binding protein
Gene Aliases: ALOX5AP; FLAP
UniProt ID: (Human) P20292
Entrez Gene ID: (Human) 241
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.