Invitrogen
Manufacturer Code:PA555874
Catalog # PIPA555874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FLAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD-synthetase; FAD1 FADS RP11-307C12.7 FAD pyrophosphorylase FAD synthase FAD synthetase FAD-synthetase FMN adenylyltransferase Fad1 flavin adenine dinucleotide synthetase homolog flavin adenine dinucleotide synthase flavin adenine dinucleotide synthetase; Fad1, flavin adenine dinucleotide synthetase, homolog; Flavin adenine dinucleotide synthase; FMN adenylyltransferase; Molybdenum cofactor biosynthesis protein-like region
Gene Aliases: FAD1; FADS; FLAD1; LSMFLAD; PP591
UniProt ID: (Human) Q8NFF5
Entrez Gene ID: (Human) 80308
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.