Invitrogen
Manufacturer Code:PA543310
Catalog # PIPA543310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FLAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD synthetase; FAD-synthetase; FAD1 flavin adenine dinucleotide synthetase homolog; Fad1, flavin adenine dinucleotide synthetase, homolog; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; FMN adenylyltransferase; Molybdenum cofactor biosynthesis protein-like region
Gene Aliases: A930017E24Rik; fa92c06; FAD1; FADS; FLAD1; LSMFLAD; PP591; wu:fa92c06; zgc:91843
UniProt ID: (Human) Q8NFF5, (Mouse) Q8R123, (Zebrafish) Q68EH8
Entrez Gene ID: (Pig) 100153896, (Horse) 100063090, (Bovine) 506682, (Human) 80308, (Dog) 490438, (Rabbit) 100346208, (Mouse) 319945, (Rat) 751787, (Zebrafish) 445492
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.