Abnova
Manufacturer Code:H00080308K
Catalog # 89-208-695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human FLAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human FLAD1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD-synthetase; FAD1 FADS MGC31803 MGC40255 PP591 RP11-307C12.7; Fad1, flavin adenine dinucleotide synthetase, homolog; Flavin adenine dinucleotide synthase; FMN adenylyltransferase; Molybdenum cofactor biosynthesis protein-like region
Gene Aliases: FAD1; FADS; FLAD1; LSMFLAD; PP591
UniProt ID: (Human) Q8NFF5
Entrez Gene ID: (Human) 80308
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.