Invitrogen
Manufacturer Code:PA552989
Catalog # PIPA552989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FKTN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fukutin; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; patient fukutin; Ribitol-5-phosphate transferase; RP11-235C23.1 CMD1X FCMD LGMD2M MDDGA4 MDDGB4 MDDGC4 Fukuyama type congenital muscular dystrophy protein patient fukutin
Gene Aliases: CMD1X; FCMD; FKTN; LGMD2M; MDDGA4; MDDGB4; MDDGC4
UniProt ID: (Human) O75072
Entrez Gene ID: (Human) 2218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.