Invitrogen
Manufacturer Code:PA542692
Catalog # PIPA542692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FKTN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fukutin; Fukuyama type congenital muscular dystrophy (fukutin); Fukuyama type congenital muscular dystrophy homolog; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; patient fukutin; Ribitol-5-phosphate transferase
Gene Aliases: CMD1X; D830030O17Rik; FCMD; FKTN; LGMD2M; MDDGA4; MDDGB4; MDDGC4
UniProt ID: (Human) O75072, (Mouse) Q8R507
Entrez Gene ID: (Human) 2218, (Dog) 481652, (Pig) 100521942, (Rabbit) 100355254, (Horse) 100054199, (Rat) 362520, (Mouse) 246179, (Guinea pig) 100722995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.