Novus Biologicals
Manufacturer Code:NBP15964020UL
Catalog # NBP15964020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to FKTN The peptide sequence was selected from the N terminal of FKTN. Peptide sequence TAFALQYHLWKNEEGWFRIAENMGFQCLKIESKDPRLDGIDSLSGTEIPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMD1X EC 2.- FCMDMGC134945 fukutin Fukuyama type congenital muscular dystrophy (fukutin) Fukuyama type congenital muscular dystrophy protein Fukuyama-type congenital muscular dystrophy protein LGMD2MMGC134944 MDDGA4 MDDGB4 MDDGC4 MGC126857 MGC138243; Fukutin; Fukuyama type congenital muscular dystrophy protein; Fukuyama-type congenital muscular dystrophy protein; patient fukutin; Ribitol-5-phosphate transferase
Gene Aliases: CMD1X; FCMD; FKTN; LGMD2M; MDDGA4; MDDGB4; MDDGC4
UniProt ID: (Human) O75072
Entrez Gene ID: (Human) 2218
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.