Proteintech
Manufacturer Code:100601AP150UL
Catalog # 50-172-5993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FKBPL Fusion Protein Ag0112 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIR1 FK506 binding protein like FKBPL NG7 WISP39; FK506-binding protein like; FK506-binding protein-like; immunophilin-like protein NG7; peptidyl-prolyl cis-trans isomerase; WAF-1/CIP1 stabilizing protein 39; WISp39
Gene Aliases: DIR1; FKBP4; FKBPL; NG7; Ppiase-X; WISP39
UniProt ID: (Human) Q9UIM3, (Mouse) O35450, (Rat) Q6MG81
Entrez Gene ID: (Human) 63943, (Mouse) 56299, (Rat) 406168
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.