Invitrogen
Manufacturer Code:PA5112904
Catalog # PIPA5112904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP8 protein (58-190AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa FK506-binding protein; 38 kDa FK506-binding protein 38 kDa FKBP 38kDa BOS_7129 FK506 binding protein 8 FK506 binding protein 8 38kDa FK506-binding protein 38 FK506-binding protein 8 FK506-binding protein 8 (38kD) FKBP prolyl isomerase 8 Fkbp38 FKBP-38 FKBP8 FKBP-8 FKBPR38 hFKBP38 mFKBP38 Peptidyl-prolyl cis-trans isomerase FKBP8 PPIase FKBP8 Rotamase Sam11; 38 kDa FKBP; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP-38; FKBP-8; FKBPR38; hFKBP38; mFKBP38; Peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; Rotamase
Gene Aliases: 38kDa; FKBP-38; FKBP-8; FKBP38; FKBP8; FKBPr38; Sam11
UniProt ID: (Human) Q14318, (Mouse) O35465
Entrez Gene ID: (Human) 23770, (Mouse) 14232
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.