Abnova
Manufacturer Code:H00002281M01
Catalog # 89-022-119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H5-1B6 |
Immunogen | FKBP1B (AAH02614 1 a.a. ~ 80 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FKBP1B.
For Research Use Only
Protein Aliases: 12.6 kDa FK506-binding protein; 12.6 kDa FKBP; calstabin 2; FK506 binding protein 1B, 12.6 kDa; FK506-binding protein 12.6; FK506-binding protein 1B; FKBP-12.6; FKBP-1B; FKBP12.6; FKBP1L; h-FKBP-12; Immunophilin FKBP12.6; OTK4; Peptidyl-prolyl cis-trans isomerase FKBP1B; PKBP1L; PPIase; PPIase FKBP1B; Rotamase
Gene Aliases: FKBP12.6; FKBP1B; FKBP1L; FKBP9; OTK4; PKBP1L; PPIase
UniProt ID: (Human) Q53TM2
Entrez Gene ID: (Human) 2281
Molecular Function: calcium-binding protein chaperone isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.