Invitrogen
Manufacturer Code:PA5109727
Catalog # PIPA5109727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-80 of human FKBP1B (NP_473374.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12.6 kDa FK506-binding protein; 12.6 kDa FK506-binding protein 12.6 kDa FKBP 12.6kDa 126 kDa FKBP AW494148 calstabin2 Calstabin-2 FK506-binding protein 1B FKBP12.6 FKBP126 FKBP-126 Fkbp1b FKBP-1B FKBP1L FKBP9 h-FKBP-12 Immunophilin FKBP12.6 OTK4 Peptidyl-prolyl cis-trans isomerase FKBP1B PKBP1L PPIase PPIase FKBP1B Rotamase; 12.6 kDa FKBP; calstabin 2; Calstabin-2; FK506 binding protein 1B, 12.6 kDa; FK506-binding protein 12.6; FK506-binding protein 1B; FKBP-12.6; FKBP-1B; h-FKBP-12; Immunophilin FKBP12.6; Peptidyl-prolyl cis-trans isomerase FKBP1B; PPIase FKBP1B; Rotamase
Gene Aliases: 12.6kDa; AW494148; calstabin2; FKBP12.6; FKBP1B; FKBP1L; FKBP9; OTK4; PKBP1L; PPIase
UniProt ID: (Human) Q53TM2, (Mouse) Q9Z2I2, (Rat) P97534
Entrez Gene ID: (Human) 2281, (Mouse) 14226, (Rat) 58950
Molecular Function: calcium-binding protein chaperone isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.