Abnova
Manufacturer Code:H00002280M01
Catalog # 89-022-118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E5-A12 |
Immunogen | FKBP1A (AAH05147 1 a.a. ~ 108 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FKBP1A.
For Research Use Only
Protein Aliases: 12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; Calstabin-1; FK506 binding protein 1A, 12kDa; FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 12; FK506-binding protein 1A; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; FKBP-12; FKBP-1A; FKBP1; FKBP12; FKBP12-Exip3; FKBP12C; Immunophilin FKBP12; Peptidyl-prolyl cis-trans isomerase FKBP1A; PKC12; PKCI2; PPIASE; PPIase FKBP1A; protein kinase C inhibitor 2; Rotamase
Gene Aliases: FKBP-12; FKBP-1A; FKBP1; FKBP12; FKBP1A; PKC12; PKCI2; PPIASE
UniProt ID: (Human) P62942
Entrez Gene ID: (Human) 2280
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.