Proteintech
Manufacturer Code:102731AP150UL
Catalog # 50-172-6169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FKBP1A Fusion Protein Ag0406 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12 kDa FK506 binding protein 12 kDa FKBP FK506 binding protein 1A FKBP 12 FKBP 1A FKBP1 FKBP12 FKBP12C FKBP1A Immunophilin FKBP12 PKC12 PKCI2 PPIASE PPIase FKBP1A Rotamase; 12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; Calstabin-1; FK506 binding protein 1A, 12kDa; FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 12; FK506-binding protein 1A; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; FKBP-1A; FKBP12-Exip3; Immunophilin FKBP12; Peptidyl-prolyl cis-trans isomerase FKBP1A; PPIase FKBP1A; protein kinase C inhibitor 2; Rotamase
Gene Aliases: Fkbp; FKBP-12; FKBP-1A; FKBP1; FKBP12; FKBP1A; PKC12; PKCI2; PPIASE
UniProt ID: (Human) P62942, (Mouse) P26883
Entrez Gene ID: (Human) 2280, (Mouse) 14225
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.