Novus Biologicals
Manufacturer Code:NBP299149AF750
Catalog # NB058465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FKBP12 (NP_463460 Met 1-Glu 108). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12 kDa FK506-binding protein; 12 kDa FKBP; 12 kDa FKBP EC 5.2.1.8 FK506 binding protein 1A 12kDa FK506-binding protein 1A FKBP1 FKBP12C FKBP12FKBP12-Exip3 FKBP-12T-cell 12-kD FKBP-1A peptidyl-prolyl cis-trans isomerase FKBP1A PPIase FKBP1A protein kinase C inhibitor 2 rotamase; calstabin 1; Calstabin-1; FK506 binding protein 1A, 12kDa; FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 12; FK506-binding protein 1A; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; FKBP-1A; FKBP12-Exip3; Immunophilin FKBP12; Peptidyl-prolyl cis-trans isomerase FKBP1A; PPIase FKBP1A; protein kinase C inhibitor 2; Rotamase
Gene Aliases: FKBP-12; FKBP-1A; FKBP1; FKBP12; FKBP1A; PKC12; PKCI2; PPIASE
UniProt ID: (Human) P62942
Entrez Gene ID: (Human) 2280
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.