Novus Biologicals
Manufacturer Code:NBP293037
Catalog # NB027969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human FKBP1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12.6 kDa FK506-binding protein; 12.6 kDa FK506-binding protein calstabin 212.6 kDa FKBP EC 5.2.1.8 FK506 binding protein 1B 12.6 kDa FK506-binding protein 12.6 FK506-binding protein 1B FK506-binding protein 1B (12.6 kD) FKBP-12.6 FKBP12.6h-FKBP-12 FKBP-1B FKBP1L FKBP9 Immunophilin FKBP12.6 OTK4PPIase FKBP1B peptidyl-prolyl cis-trans isomerase FKBP1B PKBP1L PPIase Rotamase; 12.6 kDa FKBP; calstabin 2; FK506 binding protein 1B, 12.6 kDa; FK506-binding protein 12.6; FK506-binding protein 1B; FKBP-12.6; FKBP-1B; h-FKBP-12; Immunophilin FKBP12.6; Peptidyl-prolyl cis-trans isomerase FKBP1B; PPIase FKBP1B; Rotamase
Gene Aliases: FKBP12.6; FKBP1B; FKBP1L; FKBP9; OTK4; PKBP1L; PPIase
UniProt ID: (Human) Q53TM2
Entrez Gene ID: (Human) 2281
Molecular Function:
calcium-binding protein
chaperone
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.