Invitrogen
Manufacturer Code:PA541914
Catalog # PIPA541914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FIP1L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Factor interacting with PAP; FIP1 like 1; FIP1 like 1 (S. cerevisiae) DKFZp586K0717 FLJ33619 Rhe hFip1; FIP1-like 1 protein; FIP1L1 cleavage and polyadenylation specific factor subunit; hFip1; Pre-mRNA 3'-end-processing factor FIP1; Rearranged in hypereosinophilia
Gene Aliases: 1300019H17Rik; FIP1; FIP1L1; hFip1; RHE; Rje; wu:fd36f11; zgc:103421
UniProt ID: (Human) Q6UN15, (Mouse) Q9D824, (Rat) Q5U317, (Zebrafish) Q5XJD3
Entrez Gene ID: (Dog) 612980, (Human) 81608, (Horse) 100060154, (Bovine) 100138550, (Rabbit) 100341056, (Mouse) 66899, (Rat) 289582, (Guinea pig) 100725850, (Zebrafish) 450021
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.