Novus Biologicals
Manufacturer Code:NBP297167H
Catalog # NB101163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human FIGNL1 fragment. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.6.4 EC 3.6.4.- fidgetin-like 1 fidgetin-like protein 1; fidgetin-like 1; Fidgetin-like protein 1
Gene Aliases: FIGNL1
UniProt ID: (Human) Q6PIW4
Entrez Gene ID: (Human) 63979
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.