Proteintech
Manufacturer Code:176041AP150UL
Catalog # 50-173-1680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FIGNL1 Fusion Protein Ag11709 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fidgetin like 1 Fidgetin like protein 1 FIGNL1; fidgetin-like 1; Fidgetin-like protein 1
Gene Aliases: FIGNL1; S30
UniProt ID: (Human) Q6PIW4, (Rat) Q6GX84, (Mouse) Q8BPY9
Entrez Gene ID: (Human) 63979, (Rat) 289777, (Mouse) 60530
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.