Invitrogen
Manufacturer Code:PA572041
Catalog # PIPA572041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 846-874 amino acids from the C-terminal region of human FIG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FIG4 homolog, SAC domain containing lipid phosphatase; FIG4 homolog, SAC1 domain containing lipid phosphatase; FIG4 homolog, SAC1 lipid phosphatase domain containing; Phosphatidylinositol 3,5-bisphosphate 5-phosphatase; Polyphosphoinositide phosphatase; RP1-249I4.1 ALS11 CMT4J KIAA0274 SAC3 dJ249I4.1 FIG4 homolog SAC domain containing lipid phosphatase FIG4 homolog SAC1 domain containing lipid phosphatase SAC domain-containing protein 3 Sac domain-containing inositol phosphatase 3 phosphatidylinositol 3 5-bisphosphate 5-phosphatase polyphosphoinositide phosphatase; Sac domain-containing inositol phosphatase 3; SAC domain-containing protein 3
Gene Aliases: A530089I17Rik; AI326867; ALS11; BTOP; CMT4J; dJ249I4.1; FIG4; KIAA0274; SAC3; YVS
UniProt ID: (Human) Q92562, (Mouse) Q91WF7
Entrez Gene ID: (Human) 9896, (Mouse) 103199
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.