Invitrogen
Manufacturer Code:PA554524
Catalog # PIPA554524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FHOD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FH1/FH2 domain-containing protein 3; FHOS2 Formactin2 FH1/FH2 domain-containing protein 3 formactin-2 formin homolog overexpressed in spleen 2 hFHOS2; Formactin-2; Formin homolog overexpressed in spleen 2; hFHOS2
Gene Aliases: FHOD3; FHOS2; Formactin2; KIAA1695
UniProt ID: (Human) Q2V2M9
Entrez Gene ID: (Human) 80206
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.