Invitrogen
Manufacturer Code:PA523313
Catalog # PIPA523313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (1263 QKQKRANHRERNKTRGK 1279) of FHOD3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FH1/FH2 domain-containing protein 3; FHOS2 Formactin2 FH1/FH2 domain-containing protein 3 formactin-2 formin homolog overexpressed in spleen 2 hFHOS2; Formactin-2; Formin homolog overexpressed in spleen 2; formin-family protein FHOS2; hFHOS2; mFHOS2
Gene Aliases: A930009H06Rik; FHOD3; FHOS2; Formactin2; KIAA1695; mKIAA1695
UniProt ID: (Human) Q2V2M9, (Mouse) Q76LL6
Entrez Gene ID: (Chicken) 420952, (Human) 80206, (Dog) 480157, (Horse) 100067548, (Bovine) 785433, (Mouse) 225288
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.