Novus Biologicals
Manufacturer Code:NBP224544
Catalog # NBP224544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 1250-1300 of human FHOD3 (CRA-d) was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FH1/FH2 domain-containing protein 3; FH1/FH2 domain-containing protein 3 FHOS2 formactin-2 Formactin2 formin homolog overexpressed in spleen 2 formin homology 2 domain containing 3 hFHOS2 KIAA1695; Formactin-2; Formin homolog overexpressed in spleen 2; hFHOS2
Gene Aliases: FHOD3; FHOS2; Formactin2; KIAA1695
UniProt ID: (Human) Q2V2M9
Entrez Gene ID: (Human) 80206
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.