Proteintech
Manufacturer Code:216191AP150UL
Catalog # 50-173-3515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FHL2 Fusion Protein Ag1378 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAG11 DRAL FHL 2 FHL2 four and a half LIM domains 2 LIM domain protein DRAL Skeletal muscle LIM protein 3 SLIM 3 SLIM3; aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; FHL-2; Four and a half LIM domains protein 2; LIM domain protein DRAL; Skeletal muscle LIM-protein 3; SLIM 3; SLIM-3
Gene Aliases: AAG11; DRAL; FHL-2; FHL2; SLIM-3; SLIM3
UniProt ID: (Human) Q14192, (Mouse) O70433, (Rat) O35115
Entrez Gene ID: (Human) 2274, (Mouse) 14200, (Rat) 63839
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.