Proteintech
Manufacturer Code:109911AP150UL
Catalog # 50-172-6773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FHL1 Fusion Protein Ag1448 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FHL; FHL 1 FHL1 FHL1B FLH1A four and a half LIM domains 1 KYO T Skeletal muscle LIM protein 1 SLIM SLIM 1 SLIM1 XMPMA; FHL-1; four and a half LIM domains a; Four and a half LIM domains protein 1; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KyoT; LIM protein SLIMMER; RAM14-1; RBP-associated molecule 14-1; Skeletal muscle LIM-protein 1; SLIM
Gene Aliases: cb1039; fhl; FHL-1; FHL1; FHL1A; FHL1B; fhla; FLH1A; KYOT; RAM14-1; RBMX1A; RBMX1B; SLIM; SLIM-1; SLIM1; SLIMMER; XMPMA; zgc:92025
UniProt ID: (Human) Q13642, (Mouse) P97447
Entrez Gene ID: (Human) 2273, (Mouse) 14199, (Zebrafish) 399646, (Zebrafish) 387528
Molecular Function:
RNA binding protein
actin family cytoskeletal protein
cytoskeletal protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.