Invitrogen
Manufacturer Code:MA535753
Catalog # PIMA535753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1401 |
Immunogen | A synthesized peptide derived from human FHIT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AP3A hydrolase; AP3A hydrolase AP3Aase AW045638 Bis(5&prime-adenosyl)-triphosphatase bis5&prime-adenosyl-triphosphatase Diadenosine 5&prime5&prime&prime&prime-P1P3-triphosphate hydrolase Dinucleosidetriphosphatase FHIT Fra14A2 FRA3B Fragile histidine triad protein; AP3Aase; Bis(5'-adenosyl)-triphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; Dinucleosidetriphosphatase; Fragile histidine triad protein
Gene Aliases: AP3Aase; FHIT; FRA3B
UniProt ID: (Human) P49789, (Rat) Q9JIX3
Entrez Gene ID: (Human) 2272, (Rat) 60398
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.