Invitrogen
Manufacturer Code:PA543785
Catalog # PIPA543785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FGL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: associated with MHV-3 induced prothrombinase activity; Cytotoxic T-lymphocyte-specific protein; fibrinogen-like 2; Fibrinogen-like 2 T49 pT49; Fibrinogen-like protein 2; Fibroleukin; Prothrombinase; pT49
Gene Aliases: AI385601; FGL2; Fiblp; musfiblp; pT49; T49
UniProt ID: (Bovine) Q29RY7, (Human) Q14314, (Mouse) P12804
Entrez Gene ID: (Dog) 475902, (Bovine) 511711, (Rabbit) 100351573, (Pig) 448808, (Horse) 100050110, (Human) 10875, (Mouse) 14190, (Rat) 84586
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.