Abnova
Manufacturer Code:MAB10648
Catalog # 89-108-845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H2B10B2 |
Immunogen | Recombinant protein corresponding to human FGFR4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant FGFR4.
For Research Use Only
Protein Aliases: CD334; FGFR-4; Fibroblast growth factor receptor 4; hydroxyaryl-protein kinase; JTK2; MGC20292; protein-tyrosine kinase; TKF; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Aliases: CD334; FGFR4; JTK2; TKF
UniProt ID: (Human) P22455
Entrez Gene ID: (Human) 2264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.