R&D Systems
Manufacturer Code:AF2265
Catalog # AF2265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse FGF R4 Leu19-Asp366 Accession # CAA42551 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD334; CD334 CD334 antigen EC 2.7.10 EC 2.7.10.1 FGF R4 FGFR-4 fibroblast growth factor receptor 4 JTK2hydroxyaryl-protein kinase MGC20292 protein-tyrosine kinase TKF tyrosine kinase related to fibroblast growth factor receptor tyrosylprotein kinase; FGFR-4; Fibroblast growth factor receptor 4; hydroxyaryl-protein kinase; protein-tyrosine kinase; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Aliases: CD334; FGFR4; JTK2; TKF
UniProt ID: (Human) P22455
Entrez Gene ID: (Human) 2264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.