Proteintech
Manufacturer Code:110981AP150UL
Catalog # 50-172-6874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGFR4 Fusion Protein Ag1573 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD334; CD334 FGFR 4 FGFR4 JTK2 TKF; CTLA-2-beta protein; FGFR-4; Fibroblast growth factor receptor 4; fibroblast growth factor receptor 4 16 minus form; hydroxyaryl-protein kinase; protein-tyrosine kinase; Protein-tyrosine kinase receptor MPK-11; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Aliases: CD334; Fgfr-4; FGFR4; JTK2; Mpk-11; TKF
UniProt ID: (Human) P22455, (Rat) Q498D6, (Mouse) Q03142
Entrez Gene ID: (Human) 2264, (Rat) 25114, (Mouse) 14186
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.