OriGene
Manufacturer Code:CF801052
Catalog # 50-166-5429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 23-375 of human FGFR3 produced in Insect. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACH CD333 CEK2 HSFGFR3EX JTK4; CD333; FGFR-3; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; hydroxyaryl-protein kinase; tyrosine kinase JTK4
Gene Aliases: ACH; CD333; CEK2; FGFR3; HSFGFR3EX; JTK4
UniProt ID: (Human) P22607
Entrez Gene ID: (Human) 2261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.