Invitrogen
Manufacturer Code:MA531141
Catalog # PIMA531141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 42 |
Immunogen | Recombit Mouse FGFR3/CD333 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: fibroblast growth factor receptor 3; Fibroblast Growth Factor Receptor 3 ACH CEK2 HSFGFR3EX JTK4 FGFR-3 fibroblast growth factor receptor 3 variant 4 hydroxyaryl-protein kinase tyrosine kinase JTK4 FGF Receptor 3; heparin-binding growth factor receptor
Gene Aliases: CD333; Fgfr-3; Flg-2; FR3; HBGFR; Mfr3; sam3
Entrez Gene ID: (Mouse) 14184
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.