R&D Systems
Manufacturer Code:FAB766A
Catalog # FAB766A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 136334 |
Immunogen | Mouse myeloma cell line NS0- and Sf21-derived recombinant human FGF R3 alpha (IIIb) |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD333; CD333 CEK EC 2.7.10 FGF R3 fibroblast growth factor receptor 3 HSFGFR3EX JTK4 JTK4thanatophoric dwarfism; FGFR-3; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; hydroxyaryl-protein kinase; tyrosine kinase JTK4
Gene Aliases: ACH; CD333; CEK2; FGFR3; HSFGFR3EX; JTK4
UniProt ID: (Human) P22607
Entrez Gene ID: (Human) 2261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.