Invitrogen
Manufacturer Code:PA137687
Catalog # 01-673-454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from internal region of human FGFR-3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD333; FGFR-3; Fibroblast growth factor receptor 3; Fibroblast Growth Factor Receptor 3 ACH CEK2 HSFGFR3EX JTK4 FGFR-3 fibroblast growth factor receptor 3 variant 4 hydroxyaryl-protein kinase tyrosine kinase JTK4 FGF Receptor 3; fibroblast growth factor receptor 3 variant 4; hydroxyaryl-protein kinase; tyrosine kinase JTK4
Gene Aliases: ACH; CD333; CEK2; FGFR3; HSFGFR3EX; JTK4
UniProt ID: (Human) P22607
Entrez Gene ID: (Human) 2261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.