Novus Biologicals
Manufacturer Code:NBP300287AF750
Catalog # NB091929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FGFR1OP (O95684 Ala2-Ala379). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Centrosomal protein 43; FGFR1 oncogene partner; FGFR1 oncogene partner FOPfibroblast growth factor receptor 1 oncogene partner; fibroblast growth factor receptor 1 oncogene partner
Gene Aliases: CEP43; FGFR1OP; FOP
UniProt ID: (Human) O95684
Entrez Gene ID: (Human) 11116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.