Proteintech
Manufacturer Code:113431AP150UL
Catalog # 50-172-7077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGFR1OP Fusion Protein Ag1891 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Centrosomal protein 43; FGFR1 oncogene partner; FGFR1 oncogene partner FGFR1OP FOP; fibroblast growth factor receptor 1 oncogene partner
Gene Aliases: 4930553O10Rik; CEP43; FGFR1OP; FOP
UniProt ID: (Human) O95684, (Mouse) Q66JX5, (Rat) Q4V7C1
Entrez Gene ID: (Human) 11116, (Mouse) 75296, (Rat) 683722
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.