Novus Biologicals
Manufacturer Code:NB120829
Catalog # NB120829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M2F12 |
Immunogen | Recombinant human ectodomain of FGFr1a expressed in E. coli beginning with pro23 antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Basic fibroblast growth factor receptor 1; basic fibroblast growth factor receptor 1 BFGFR bFGF-R-1 CD331 CD331 antigen CEK EC 2.7.10 EC 2.7.10.1 FGFBR FGFR-1 fibroblast growth factor receptor 1 FLGH3 FLJ99988 FLT-2 FLT2H4 Fms-like tyrosine kinase 2 fms-related tyrosine kinase 2 H2 H5 HBGFR heparin-binding growth factor receptor hydroxyaryl-protein kinase KAL 2 KAL2 N-SAM OGD Proto-oncogene c-Fgr soluble FGFR1 variant 1 soluble FGFR1 variant 2; BFGFR; CD331; FGFR-1; FGFR1/PLAG1 fusion; Fibroblast growth factor receptor 1; FLT-2; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; heparin-binding growth factor receptor; hydroxyaryl-protein kinase; N-sam; Proto-oncogene c-Fgr
Gene Aliases: bFGF-R-1; BFGFR; CD331; CEK; ECCL; FGFBR; FGFR-1; FGFR1; FLG; FLT-2; FLT2; HBGFR; HH2; HRTFDS; KAL2; N-SAM; OGD
UniProt ID: (Human) P11362
Entrez Gene ID: (Human) 2260
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.