Proteintech
Manufacturer Code:265541AP150UL
Catalog # 50-173-5374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGF9 Fusion Protein Ag24244 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF 9 FGF9 Fibroblast growth factor 9 GAF Glia activating factor HBFG 9 HBGF 9; FGF-9; Fibroblast growth factor 9; fibroblast growth factor 9 (glia-activating factor); GAF; Glia-activating factor; HBGF-9; Heparin-binding growth factor 9
Gene Aliases: FGF-9; FGF9; GAF; HBFG-9; HBGF-9; SYNS3
UniProt ID: (Human) P31371
Entrez Gene ID: (Human) 2254
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.