Invitrogen
Manufacturer Code:PA547598
Catalog # PIPA547598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse FGF-8b Gln23-Arg215 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGF; androgen-induced AIGF HBGF-8 MGC149376 androgen-induced growth factor fibroblast growth factor 8; Androgen-induced growth factor; FGF-8; Fibroblast growth factor 8; HBGF-8; Heparin-binding growth factor 8
Gene Aliases: Aigf; Fgf-8; Fgf8
UniProt ID: (Mouse) P37237
Entrez Gene ID: (Mouse) 14179
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.