Invitrogen
Manufacturer Code:PA5114734
Catalog # PIPA5114734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FGF8 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGF; AIGF androgen-induced Androgen-induced growth factor FGF FGF8 FGF-8 Fibroblast growth factor fibroblast growth factor 8 fibroblast growth factor 8 (androgen-induced) HBGF-8 heparin-binding growth factor 8 HH6 KAL6 MGC149376; Androgen-induced growth factor; FGF-8; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8
Gene Aliases: AIGF; FGF-8; FGF8; HBGF-8; HH6; KAL6
UniProt ID: (Human) P55075, (Mouse) P37237
Entrez Gene ID: (Human) 2253, (Rat) 29349, (Mouse) 14179
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.