Invitrogen
Manufacturer Code:PA569161
Catalog # PIPA569161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat FGF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-3; Fibroblast growth factor 3; fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog); HBGF-3; HBGF-3 INT2 FGF-3 INT-2 proto-oncogene protein V-INT2 murine mammary tumor virus integration site oncogene homolog fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog) heparin-binding growth factor 3 murine mammary tumor; Heparin-binding growth factor 3; INT-2 proto-oncogene protein; interacting gene 2; lia; murine mammary tumor virus integration site 2, mouse; oncogene INT2; Proto-oncogene Int-2; V-INT2 murine mammary tumor virus integration site oncogene homolog
Gene Aliases: FGF-3; FGF3; HBGF-3; id:ibd5006; Int-2; Int-P; INT2
UniProt ID: (Human) P11487, (Zebrafish) P48802
Entrez Gene ID: (Human) 2248, (Dog) 611707, (Horse) 100146649, (Mouse) 14174, (Rat) 170633, (Guinea pig) 100716446, (Zebrafish) 30549
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.