Invitrogen
Manufacturer Code:PA595597
Catalog # PIPA595597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived rat FGF23 recombit protein (Position: Y25-V251). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-23; Fibroblast growth factor 23; Fibroblast growth factor 23 FGF-23 Fgf23
Gene Aliases: Fgf23
UniProt ID: (Mouse) Q9EPC2, (Rat) Q8VI82
Entrez Gene ID: (Mouse) 64654, (Rat) 170583
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.