Invitrogen
Manufacturer Code:PA5114442
Catalog # PIPA5114442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse FGF23 recombinant protein (Position: R48-V251). Mouse FGF23 shares 71.1% and 94.6% amino acid (aa) sequence identity with human and rat FGF23 respectively. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADHR FGF Fgf23 FGF-23 FGFN Fibroblast growth factor Fibroblast growth factor 23 Fibroblast growth factor 23 C-terminal peptide Fibroblast growth factor 23 N-terminal peptide HPDR2 HYPF Phosphatonin PHPTC tumor-derived hypophosphatemia inducing factor Tumor-derived hypophosphatemia-inducing factor UNQ3027/PRO9828; FGF-23; Fibroblast growth factor 23
Gene Aliases: Fgf23
UniProt ID: (Mouse) Q9EPC2
Entrez Gene ID: (Mouse) 64654
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.