OriGene
Manufacturer Code:TA500013
Catalog # 50-167-1321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 10-155 of human bFGF produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; BFGF FGF-2 FGFB HBGF-2; FGF-2; Fibroblast growth factor 2; fibroblast growth factor 2 (basic); HBGF-2; Heparin-binding growth factor 2; prostatropin
Gene Aliases: BFGF; FGF-2; FGF2; FGFB; HBGF-2
UniProt ID: (Human) P09038
Entrez Gene ID: (Human) 2247
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.