Proteintech
Manufacturer Code:253141AP150UL
Catalog # 50-173-4853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGF17 Fusion Protein Ag17807 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF 13 FGF 17 FGF17 fibroblast growth factor 17; FGF-17; Fibroblast growth factor 17
Gene Aliases: FGF-13; FGF-17; FGF17; HH20; UNQ161/PRO187
UniProt ID: (Human) O60258, (Mouse) P63075
Entrez Gene ID: (Human) 8822, (Mouse) 14171
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.