Invitrogen
Manufacturer Code:MA527705
Catalog # PIMA527705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S235-22 |
Immunogen | Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100&percnt identical to rat 94&percnt identical to mouse. >80&percnt identity with FGF12A/FHF1A FGF14A/FHF4A and FGF11A/FHF3A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-13; FGF-13 FHF-2 FGF13 FHF2 Acidic fibroblast growth factor AFGF Beta endothelial cell growth factor Fibroblast growth factor homologous factor 2A Fibroblast growth factor 13A FGF13A Beta-endothelial cell growth factor ECGF ECGFA ECGFB Endothelial Cell Growth Factor alpha Endothelial Cell Growth Factor alpha Endothelial Cell Growth Factor beta FGF 1 FGF alpha FGFA Fibroblast Growth Factor 1 Acidic Fibroblast growth factor 1 GLIO703 HBGF 1 HBGF1 Heparin binding growth factor 1 Heparin binding growth factor 1 precursor Heparin-binding growth factor 1; FHF-2; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2
Gene Aliases: FGF-13; FGF13; FGF2; FHF-2; FHF2
UniProt ID: (Human) Q92913, (Rat) Q9ERW3, (Mouse) P70377
Entrez Gene ID: (Human) 2258, (Rat) 84488, (Mouse) 14168
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.