OriGene
Manufacturer Code:CF807761
Catalog # 50-166-8466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7E8 |
Immunogen | Full length human recombit protein of human FGF13 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-13; FGF-13 FGF2 FHF-2 FHF2; FHF-2; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2
Gene Aliases: FGF-13; FGF13; FGF2; FHF-2; FHF2
UniProt ID: (Human) Q92913
Entrez Gene ID: (Human) 2258
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.