Invitrogen
Manufacturer Code:PA544342
Catalog # PIPA544342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FGF13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-13; FHF-2; Fibroblast growth factor 13; Fibroblast growth factor 13 FGF2 FHF2 FHF-2 FGF-13; Fibroblast growth factor homologous factor 2
Gene Aliases: FGF-13; FGF13; FGF2; FHF-2; FHF2
UniProt ID: (Human) Q92913, (Rat) Q9ERW3, (Mouse) P70377
Entrez Gene ID: (Bovine) 506924, (Sheep) 101101956, (Human) 2258, (Horse) 100054495, (Dog) 492174, (Rabbit) 100343850, (Rat) 84488, (Guinea pig) 100732809, (Mouse) 14168
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.