Invitrogen
Manufacturer Code:PA527302
Catalog # PIPA527302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 176 of Human FGF13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-13; FGF-13 FGF2 FHF-2 FHF2 fibroblast growth factor homologous factor 2; FHF-2; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2
Gene Aliases: FGF-13; FGF13; FGF2; FHF-2; FHF2
UniProt ID: (Human) Q92913, (Rat) Q9ERW3, (Mouse) P70377
Entrez Gene ID: (Human) 2258, (Rat) 84488, (Mouse) 14168
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.