Proteintech
Manufacturer Code:137841AP150UL
Catalog # 50-172-9004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FGF12 Fusion Protein Ag4797 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF 12 FGF12 FGF12B FHF 1 FHF1 fibroblast growth factor 12 Myocyte activating factor; FGF-12; FHF-1; Fibroblast growth factor 12; fibroblast growth factor 12B; fibroblast growth factor FGF-12b; Fibroblast growth factor homologous factor 1; Myocyte-activating factor
Gene Aliases: AV114868; B230343J05Rik; FGF-12; FGF12; FGF12B; FHF-1; FHF1
UniProt ID: (Human) P61328, (Mouse) P61329
Entrez Gene ID: (Human) 2257, (Mouse) 14167
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.