Invitrogen
Manufacturer Code:MA535690
Catalog # PIMA535690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1414 |
Immunogen | A synthesized peptide derived from human FGF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Acidic eye-derived growth factor II Acidic fibroblast growth factor AFGF BOS_8269 Dffrx ECGF ECGFA ECGFB ECGF-beta EDGF II Endothelial cell growth factor Endothelial cell growth factor alpha Endothelial cell growth factor beta Fam FGF FGF1 Fgf-1 FGFA FGF-alpha Fibroblast growth factor Fibroblast growth factor 1 GLIO703 HBGF1 HBGF-1 Heparin-binding growth factor 1 H-FGF-a M-FGF-a Prostatropin; Acidic fibroblast growth factor; aFGF; FGF-1; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); HBGF-1; Heparin-binding growth factor 1
Gene Aliases: Dffrx; Fam; FGF-1; Fgf1; Fgfa; HBGF-1; HBGF1
UniProt ID: (Rat) P61149, (Mouse) P61148
Entrez Gene ID: (Rat) 25317, (Mouse) 14164
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.